accessibility menu, dialog, popup

UserName

CAS RN 4104-45-4

IUPAC Name: 3-(methylsulfanyl)propan-1-aminium
Synonyms: 3-methylthiopropanamine 3-methylthiopropylaminium(1+)
Molecular Weight (g/mol): 106.21
Molecular Formula: C4H12NS
InChi Key: KKYSBGWCYXYOHA-UHFFFAOYSA-O
SMILES: CSCCC[NH3+]
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)