Learn More
3-(Methylthio)propyl isothiocyanate, 98%, Thermo Scientific Chemicals
$93.40 - $93.40
Chemical Identifiers
| CAS | 505-79-3 |
|---|---|
| Molecular Formula | C5H9NS2 |
| Molecular Weight (g/mol) | 147.254 |
| MDL Number | MFCD00209521 |
| InChI Key | LDKSCZJUIURGMW-UHFFFAOYSA-N |
| Synonym | 3-methylthio propyl isothiocyanate, iberverin, 3-methylthiopropyl isothiocyanate, ibervirin, propane, 1-isothiocyanato-3-methylthio, unii-ytt13bn94z, 3-methylmercaptopropyl isothiocyanate, isothiocyanic acid, 3-methylthio propyl ester, 1-isothiocyanato-3-methylthio propane, fema no. 3312 |
| PubChem CID | 62351 |
| IUPAC Name | 1-isothiocyanato-3-methylsulfanylpropane |
| SMILES | CSCCCN=C=S |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2215009
|
Thermo Scientific Chemicals
B2215009 |
10 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 505-79-3 | |
| 147.254 | |
| LDKSCZJUIURGMW-UHFFFAOYSA-N | |
| 62351 | |
| CSCCCN=C=S |
| C5H9NS2 | |
| MFCD00209521 | |
| 3-methylthio propyl isothiocyanate, iberverin, 3-methylthiopropyl isothiocyanate, ibervirin, propane, 1-isothiocyanato-3-methylthio, unii-ytt13bn94z, 3-methylmercaptopropyl isothiocyanate, isothiocyanic acid, 3-methylthio propyl ester, 1-isothiocyanato-3-methylthio propane, fema no. 3312 | |
| 1-isothiocyanato-3-methylsulfanylpropane |
Specifications
| 505-79-3 | |
| 254°C | |
| Pungent | |
| 1.564 | |
| 10 g | |
| Moisture sensitive | |
| LDKSCZJUIURGMW-UHFFFAOYSA-N | |
| 1-isothiocyanato-3-methylsulfanylpropane | |
| 62351 | |
| 98% |
| 1.113 | |
| 110°C (230°F) | |
| C5H9NS2 | |
| MFCD00209521 | |
| UN2922 | |
| 3-methylthio propyl isothiocyanate, iberverin, 3-methylthiopropyl isothiocyanate, ibervirin, propane, 1-isothiocyanato-3-methylthio, unii-ytt13bn94z, 3-methylmercaptopropyl isothiocyanate, isothiocyanic acid, 3-methylthio propyl ester, 1-isothiocyanato-3-methylthio propane, fema no. 3312 | |
| CSCCCN=C=S | |
| 147.254 | |
| 147.25 | |
| 3-(Methylthio)propyl isothiocyanate |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 208-020-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only