accessibility menu, dialog, popup

UserName

CAS RN 505-79-3

IUPAC Name: 1-isothiocyanato-3-(methylsulfanyl)propane
Synonyms: 1-isothiocyanato-3-(methylsulfanyl)propane
Molecular Weight (g/mol): 147.25
Molecular Formula: C5H9NS2
InChi Key: LDKSCZJUIURGMW-UHFFFAOYSA-N
SMILES: CSCCCN=C=S
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)