missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)propyl Isothiocyanate 97.0+%, TCI America™

$100.11 - $100.11

Chemical Identifiers

CAS 505-79-3
Molecular Formula C5H9NS2
Molecular Weight (g/mol) 147.254
MDL Number MFCD00209521
InChI Key LDKSCZJUIURGMW-UHFFFAOYSA-N
Synonym 3-methylthio propyl isothiocyanate, iberverin, 3-methylthiopropyl isothiocyanate, ibervirin, propane, 1-isothiocyanato-3-methylthio, unii-ytt13bn94z, 3-methylmercaptopropyl isothiocyanate, isothiocyanic acid, 3-methylthio propyl ester, 1-isothiocyanato-3-methylthio propane, fema no. 3312
PubChem CID 62351
IUPAC Name 1-isothiocyanato-3-methylsulfanylpropane
SMILES CSCCCN=C=S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M20275G
View Documents
TCI America
M20275G
5 g
Each for $100.11
Only null left
 
Description

Description

Specifications

Chemical Identifiers

505-79-3
147.254
LDKSCZJUIURGMW-UHFFFAOYSA-N
62351
CSCCCN=C=S
C5H9NS2
MFCD00209521
3-methylthio propyl isothiocyanate, iberverin, 3-methylthiopropyl isothiocyanate, ibervirin, propane, 1-isothiocyanato-3-methylthio, unii-ytt13bn94z, 3-methylmercaptopropyl isothiocyanate, isothiocyanic acid, 3-methylthio propyl ester, 1-isothiocyanato-3-methylthio propane, fema no. 3312
1-isothiocyanato-3-methylsulfanylpropane

Specifications

505-79-3
122°C
MFCD00209521
3-methylthio propyl isothiocyanate, iberverin, 3-methylthiopropyl isothiocyanate, ibervirin, propane, 1-isothiocyanato-3-methylthio, unii-ytt13bn94z, 3-methylmercaptopropyl isothiocyanate, isothiocyanic acid, 3-methylthio propyl ester, 1-isothiocyanato-3-methylthio propane, fema no. 3312
CSCCCN=C=S
147.254
147.25
Liquid
Yellow
C5H9NS2
5 g
LDKSCZJUIURGMW-UHFFFAOYSA-N
1-isothiocyanato-3-methylsulfanylpropane
62351
≥97.0% (GC)
3-(Methylthio)propyl Isothiocyanate
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'