missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)propyl Isothiocyanate 97.0+%, TCI America™

Catalog No. M20275G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M20275G Supplier TCI America Supplier No. M20275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 505-79-3
Molecular Formula C5H9NS2
Molecular Weight (g/mol) 147.254
MDL Number MFCD00209521
InChI Key LDKSCZJUIURGMW-UHFFFAOYSA-N
Synonym 3-methylthio propyl isothiocyanate, iberverin, 3-methylthiopropyl isothiocyanate, ibervirin, propane, 1-isothiocyanato-3-methylthio, unii-ytt13bn94z, 3-methylmercaptopropyl isothiocyanate, isothiocyanic acid, 3-methylthio propyl ester, 1-isothiocyanato-3-methylthio propane, fema no. 3312
PubChem CID 62351
IUPAC Name 1-isothiocyanato-3-methylsulfanylpropane
SMILES CSCCCN=C=S

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 g
Formula Weight 147.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Methylthio)propyl Isothiocyanate