missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M17935G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M17935G Supplier TCI America Supplier No. M17935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 128312-11-8
Molecular Formula C7H9BO2S
Molecular Weight (g/mol) 168.017
MDL Number MFCD01319105
InChI Key TYVPOLHSKGEXIH-UHFFFAOYSA-N
Synonym 3-methylthio phenylboronic acid, 3-methylthio phenyl boronic acid, 3-meththio phenylboronic acid, 3-methylthiophenylboronic acid, 3-methylthio benzeneboronic acid, 3-methylsulfanyl phenylboronic acid, 3-boronothioanisole, 3-methylthiobenzeneboronic acid, 4-methylthiophenyl boronic acid, 3-methylthio phenyl-boronic acid
PubChem CID 2734383
IUPAC Name (3-methylsulfanylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)SC)(O)O

Specifications

Melting Point 162°C
Quantity 5 g
Formula Weight 168.02
Physical Form Crystalline Powder
Chemical Name or Material 3-(Methylthio)phenylboronic Acid (contains varying amounts of Anhydride)