accessibility menu, dialog, popup

UserName

BSCH3HOOH

CAS RN 128312-11-8

IUPAC Name: [3-(methylsulfanyl)phenyl]boronic acid
Synonyms: 3-(methylsulfanyl)phenylboronic acid
Molecular Weight (g/mol): 168.02
Molecular Formula: C7H9BO2S
InChi Key: TYVPOLHSKGEXIH-UHFFFAOYSA-N
SMILES: CSC1=CC=CC(=C1)B(O)O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results