missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthio-1-propylamine, 97%

Catalog No. AAH2584603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2584603 1 g
AAH2584606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2584603 Supplier Thermo Scientific Chemicals Supplier No. H2584603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4104-45-4 | C4H11NS | 105.199 g/mol

3-Aminopropyl Methyl Sulfideis used in cheese and seafood flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminopropyl Methyl Sulfideis used in cheese and seafood flavors.

Solubility
Insoluble in water. Soluble in ethanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Chemical Identifiers

CAS 4104-45-4
Molecular Formula C4H11NS
Molecular Weight (g/mol) 105.199
MDL Number MFCD00041898
InChI Key KKYSBGWCYXYOHA-UHFFFAOYSA-N
Synonym 3-methylthiopropylamine, 3-methylthio propylamine, 1-propanamine, 3-methylthio, 3-methylthiopropanamine, methylmercaptopropylamine, unii-1iap0cgd5s, 3-methylmercaptopropylamine, 3-methylthio-1-propanamine, 3-methylsulfanyl propan-1-amine, 1iap0cgd5s
PubChem CID 77743
ChEBI CHEBI:16696
IUPAC Name 3-methylsulfanylpropan-1-amine
SMILES CSCCCN

Specifications

Density 0.938
Boiling Point 169°C
Flash Point 62°C (144°F)
Odor Stench
Refractive Index 1.494
Linear Formula CH3S(CH2)3NH2
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Insoluble in water. Soluble in ethanol.
Formula Weight 105.2
Percent Purity 97%
Chemical Name or Material 3-Methylthio-1-propylamine
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 223-875-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only