missing translation for 'onlineSavingsMsg'
Learn More

3-Methylstyrene, 95%

Catalog No. AC450870010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450870010 1 g
AC450870050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450870010 Supplier Thermo Scientific Chemicals Supplier No. 450870010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 100-80-1 | C9H10 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-80-1
Molecular Formula C9H10
Molecular Weight (g/mol) 118.18
InChI Key JZHGRUMIRATHIU-UHFFFAOYSA-N
Synonym 3-methylstyrene, 3-vinyltoluene, 1-methyl-3-vinylbenzene, m-vinyltoluene, m-methylstyrene, benzene, 1-ethenyl-3-methyl, styrene, m-methyl, m-methyl styrene, meta-methylstyrene, unii-ffk4i9430g
PubChem CID 7529
IUPAC Name 1-ethenyl-3-methylbenzene
SMILES CC1=CC=CC(=C1)C=C

Specifications

Melting Point -82°C to 81°C
Boiling Point 170.0°C to 171.0°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 118.18
Percent Purity 95%
Chemical Name or Material 3-Methylstyrene
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 202-889-2

RUO – Research Use Only