missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylcyclohexanone 97.0+%, TCI America™
Supplier: TCI America M0198100ML
Specifications
| 3-Methylcyclohexanone | |
| Yellow | |
| C7H12O | |
| 100 mL | |
| 3-methylcyclohexanone, cyclohexanone, 3-methyl, methyl-3 cyclohexanone-1, 3-methyl cyclohexanone, methyl-3 cyclohexanone-1 french, m-methylcyclohexanone, r-3-methylcyclohexanone, +-3r-3-methylcyclohexanone, cyclohexanone, 3-methyl-, r, cyclohexanone, 3-methyl-, s | |
| CC1CCCC(=O)C1 | |
| 112.17 | |
| 112.17 | |
| Liquid |
| 591-24-2 | |
| 169°C | |
| MFCD00001639 | |
| 2297 | |
| UJBOOUHRTQVGRU-UHFFFAOYNA-N | |
| 3-methylcyclohexan-1-one | |
| 11567 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 591-24-2 | |
| 112.17 | |
| UJBOOUHRTQVGRU-UHFFFAOYNA-N | |
| 11567 | |
| CC1CCCC(=O)C1 |
| C7H12O | |
| MFCD00001639 | |
| 3-methylcyclohexanone, cyclohexanone, 3-methyl, methyl-3 cyclohexanone-1, 3-methyl cyclohexanone, methyl-3 cyclohexanone-1 french, m-methylcyclohexanone, r-3-methylcyclohexanone, +-3r-3-methylcyclohexanone, cyclohexanone, 3-methyl-, r, cyclohexanone, 3-methyl-, s | |
| 3-methylcyclohexan-1-one |
Safety and Handling
EINECSNumber : (3)-2411
RTECSNumber : GW1750100
TSCA : Yes