accessibility menu, dialog, popup

UserName

CH3O

CAS RN 591-24-2

IUPAC Name: 3-methylcyclohexan-1-one
Synonyms: 3-methylcyclohexanone
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: UJBOOUHRTQVGRU-UHFFFAOYNA-N
SMILES: CC1CCCC(=O)C1
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)