missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcyclohexanone 97.0+%, TCI America™

Catalog No. p-7132049
Change view
Click to view available options
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
M019825ML 25 mL
M0198100ML 100 mL
M0198500ML 500 mL
3 options

Catalog No. M019825ML

Supplier: TCI America M019825ML

Only null left

Chemical Identifiers

CAS 591-24-2
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00001639
InChI Key UJBOOUHRTQVGRU-UHFFFAOYNA-N
Synonym 3-methylcyclohexanone, cyclohexanone, 3-methyl, methyl-3 cyclohexanone-1, 3-methyl cyclohexanone, methyl-3 cyclohexanone-1 french, m-methylcyclohexanone, r-3-methylcyclohexanone, +-3r-3-methylcyclohexanone, cyclohexanone, 3-methyl-, r, cyclohexanone, 3-methyl-, s
PubChem CID 11567
IUPAC Name 3-methylcyclohexan-1-one
SMILES CC1CCCC(=O)C1

Specifications

Color Yellow
Boiling Point 169°C
Quantity 25 mL
UN Number 2297
Formula Weight 112.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylcyclohexanone
EINECSNumber (3)-2411
RTECSNumber GW1750100
TSCA Yes