missing translation for 'onlineSavingsMsg'
Learn More

3-Methylanisole, 99%

Catalog No. AAB2145518
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2145518 50 g
AAB2145530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2145518 Supplier Thermo Scientific Chemicals Supplier No. B2145518
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100-84-5 | C8H10O | 122.17 g/mol

3-Methylanisole, is widely used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photo initiators and agrochemicals. It is also used as a solvent and in perfumery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylanisole, is widely used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photo initiators and agrochemicals. It is also used as a solvent and in perfumery.

Solubility
Soluble in alcohol and insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 100-84-5
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00008395
InChI Key OSIGJGFTADMDOB-UHFFFAOYSA-N
Synonym 3-methylanisole, 3-methoxytoluene, m-methylanisole, m-cresol methyl ether, m-methoxytoluene, benzene, 1-methoxy-3-methyl, anisole, m-methyl, methyl m-tolyl ether, m-cresyl methyl ether, methyl m-cresyl ether
PubChem CID 7530
IUPAC Name 1-methoxy-3-methylbenzene
SMILES COC1=CC=CC(C)=C1

Specifications

Density 0.969
Boiling Point 175°C to 176°C
Flash Point 54°C (129°F)
Odor Pleasant
Refractive Index 1.513
Quantity 50 g
UN Number UN3271
Beilstein 907023
Solubility Information Soluble in alcohol and insoluble in water.
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material 3-Methylanisole
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 202-893-4
RTECSNumber BZ8778000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only