accessibility menu, dialog, popup

UserName

CAS RN 100-84-5

IUPAC Name: 1-methoxy-3-methylbenzene
Synonyms: M-methylanisole
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: OSIGJGFTADMDOB-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)