missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-N-methylbenzylamine 97.0+%, TCI America™
$127.07 - $371.56
Chemical Identifiers
| CAS | 39180-84-2 |
|---|---|
| Molecular Formula | C9H13N |
| Molecular Weight (g/mol) | 135.21 |
| MDL Number | MFCD05865181 |
| InChI Key | WCTNVGNEUDTSOZ-UHFFFAOYSA-N |
| Synonym | 3-methyl-n-methylbenzylamine, n-methyl-1-m-tolyl methanamine, 3-methylbenzyl methylamine, methyl 3-methylphenyl methyl amine, benzenemethanamine, n,3-dimethyl, acmc-20a7fx, n,3-dimethylbenzylamine, methyl 3-methylbenzyl amine, n-methyl-3-methylbenzylamine, ksc493m5b |
| PubChem CID | 13794465 |
| IUPAC Name | N-methyl-1-(3-methylphenyl)methanamine |
| SMILES | CC1=CC(=CC=C1)CNC |
Chemical Identifiers
| 39180-84-2 | |
| 135.21 | |
| WCTNVGNEUDTSOZ-UHFFFAOYSA-N | |
| 13794465 | |
| CC1=CC(=CC=C1)CNC |
| C9H13N | |
| MFCD05865181 | |
| 3-methyl-n-methylbenzylamine, n-methyl-1-m-tolyl methanamine, 3-methylbenzyl methylamine, methyl 3-methylphenyl methyl amine, benzenemethanamine, n,3-dimethyl, acmc-20a7fx, n,3-dimethylbenzylamine, methyl 3-methylbenzyl amine, n-methyl-3-methylbenzylamine, ksc493m5b | |
| N-methyl-1-(3-methylphenyl)methanamine |
Specifications
| 39180-84-2 | |
| 92°C | |
| MFCD05865181 | |
| 2735 | |
| WCTNVGNEUDTSOZ-UHFFFAOYSA-N | |
| N-methyl-1-(3-methylphenyl)methanamine | |
| 13794465 | |
| ≥97.0% (GC) | |
| 3-Methyl-N-methylbenzylamine |
| Yellow | |
| C9H13N | |
| 1 mL | |
| 3-methyl-n-methylbenzylamine, n-methyl-1-m-tolyl methanamine, 3-methylbenzyl methylamine, methyl 3-methylphenyl methyl amine, benzenemethanamine, n,3-dimethyl, acmc-20a7fx, n,3-dimethylbenzylamine, methyl 3-methylbenzyl amine, n-methyl-3-methylbenzylamine, ksc493m5b | |
| CC1=CC(=CC=C1)CNC | |
| 135.21 | |
| 135.21 | |
| Liquid |
Safety and Handling
TSCA : No