CAS RN 39180-84-2
CAS RN 39180-84-2
IUPAC Name:
methyl[(3-methylphenyl)methyl]amine
Synonyms:
methyl[(3-methylphenyl)methyl]amine
Molecular Weight (g/mol):
135.21
Molecular Formula:
C9H13N
InChi Key:
WCTNVGNEUDTSOZ-UHFFFAOYSA-N
SMILES:
CNCC1=CC=CC(C)=C1
1
–
1
of
1
results
1
3-Methyl-N-methylbenzylamine 97.0+%, TCI America™
CAS: 39180-84-2 Molecular Formula: C9H13N Molecular Weight (g/mol): 135.21 MDL Number: MFCD05865181 InChI Key: WCTNVGNEUDTSOZ-UHFFFAOYSA-N Synonym: 3-methyl-n-methylbenzylamine,n-methyl-1-m-tolyl methanamine,3-methylbenzyl methylamine,methyl 3-methylphenyl methyl amine,benzenemethanamine, n,3-dimethyl,acmc-20a7fx,n,3-dimethylbenzylamine,methyl 3-methylbenzyl amine,n-methyl-3-methylbenzylamine,ksc493m5b PubChem CID: 13794465 IUPAC Name: N-methyl-1-(3-methylphenyl)methanamine SMILES: CC1=CC(=CC=C1)CNC