missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-N-methylbenzylamine 97.0+%, TCI America™

Catalog No. M28245ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28245ML 5 mL
M28241ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28245ML Supplier TCI America Supplier No. M28245ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39180-84-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD05865181
InChI Key WCTNVGNEUDTSOZ-UHFFFAOYSA-N
Synonym 3-methyl-n-methylbenzylamine, n-methyl-1-m-tolyl methanamine, 3-methylbenzyl methylamine, methyl 3-methylphenyl methyl amine, benzenemethanamine, n,3-dimethyl, acmc-20a7fx, n,3-dimethylbenzylamine, methyl 3-methylbenzyl amine, n-methyl-3-methylbenzylamine, ksc493m5b
PubChem CID 13794465
IUPAC Name N-methyl-1-(3-methylphenyl)methanamine
SMILES CC1=CC(=CC=C1)CNC

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 mL
UN Number 2735
Formula Weight 135.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-N-methylbenzylamine