missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(3-pyrrolidinyl)-1,2,4-oxadiazole hydrochloride

CAS: 1121057-52-0 | C7H12ClN3O | 189.643 g/mol

Supplier:  Thermo Scientific Chemicals H5166406

 View more versions of this product

Catalog No. AAH5166406


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Methyl-5-(3-pyrrolidinyl)-1,2,4-oxadiazole hydrochloride
C7H12ClN3O
5 g
3-methyl-5-pyrrolidin-3-yl-1,2,4-oxadiazole hydrochloride, 3-methyl-5-3-pyrrolidinyl-1,2,4-oxadiazole hydrochloride, 3-methyl-5-3-pyrrolidinyl-1,2,4-oxadiazole hcl
CC1=NOC(=N1)C2CCNC2.Cl
189.643
189.65
1121057-52-0
MFCD11870728
Air Sensitive
ZNFSKFKUBLQDHA-UHFFFAOYSA-N
3-methyl-5-pyrrolidin-3-yl-1,2,4-oxadiazole;hydrochloride
53400826

Chemical Identifiers

1121057-52-0
189.643
ZNFSKFKUBLQDHA-UHFFFAOYSA-N
53400826
CC1=NOC(=N1)C2CCNC2.Cl
C7H12ClN3O
MFCD11870728
3-methyl-5-pyrrolidin-3-yl-1,2,4-oxadiazole hydrochloride, 3-methyl-5-3-pyrrolidinyl-1,2,4-oxadiazole hydrochloride, 3-methyl-5-3-pyrrolidinyl-1,2,4-oxadiazole hcl
3-methyl-5-pyrrolidin-3-yl-1,2,4-oxadiazole;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only