missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(3-pyrrolidinyl)-1,2,4-oxadiazole hydrochloride

Catalog No. p-7034019
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51664MD 250 mg
AAH5166403 1 g
AAH5166406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH51664MD Supplier Thermo Scientific Chemicals Supplier No. H51664MD

CAS: 1121057-52-0 | C7H12ClN3O | 189.643 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1121057-52-0
Molecular Formula C7H12ClN3O
Molecular Weight (g/mol) 189.643
MDL Number MFCD11870728
InChI Key ZNFSKFKUBLQDHA-UHFFFAOYSA-N
Synonym 3-methyl-5-pyrrolidin-3-yl-1,2,4-oxadiazole hydrochloride, 3-methyl-5-3-pyrrolidinyl-1,2,4-oxadiazole hydrochloride, 3-methyl-5-3-pyrrolidinyl-1,2,4-oxadiazole hcl
PubChem CID 53400826
IUPAC Name 3-methyl-5-pyrrolidin-3-yl-1,2,4-oxadiazole;hydrochloride
SMILES CC1=NOC(=N1)C2CCNC2.Cl

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 189.65
Chemical Name or Material 3-Methyl-5-(3-pyrrolidinyl)-1,2,4-oxadiazole hydrochloride
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only