accessibility menu, dialog, popup

UserName

CAS RN 1121057-52-0

IUPAC Name: 3-methyl-5-(pyrrolidin-3-yl)-1,2,4-oxadiazole hydrochloride
Synonyms: 3-methyl-5-(pyrrolidin-3-yl)-1,2,4-oxadiazole hydrochloride
Molecular Weight (g/mol): 189.64
Molecular Formula: C7H12ClN3O
InChi Key: ZNFSKFKUBLQDHA-UHFFFAOYNA-N
SMILES: Cl.CC1=NOC(=N1)C1CCNC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)