missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitrobenzonitrile, 97%

Catalog No. AAA1130114
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1130114 25 g
AAA1130106 5 g
AAA1130109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1130114 Supplier Thermo Scientific Chemicals Supplier No. A1130114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96784-54-2 | C8H6N2O2 | 162.148 g/mol

3-Methyl-4-nitrobenzonitrile is used to produce 4-amino-3-methyl-benzonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-4-nitrobenzonitrile is used to produce 4-amino-3-methyl-benzonitrile.

Solubility
Insoluble in water

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 96784-54-2
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00017615
InChI Key IHVNKSXTJZNBQA-UHFFFAOYSA-N
Synonym benzonitrile, 3-methyl-4-nitro, 5-cyano-2-nitrotoluene, 3-methyl-4-nitro-benzonitrile, 3-methyl-4-nitrobenzenecarbonitrile, 4-nitro-m-tolunitrile, 4-cyano-2-methylnitrobenzene, ihvnksxtjznbqa-uhfffaoysa
PubChem CID 2801434
IUPAC Name 3-methyl-4-nitrobenzonitrile
SMILES CC1=C(C=CC(=C1)C#N)[N+](=O)[O-]

Specifications

Melting Point 78°C to 84°C
Quantity 25 g
UN Number UN3439
Beilstein 2443778
Solubility Information Insoluble in water
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-nitrobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only