accessibility menu, dialog, popup

UserName

CAS RN 96784-54-2

IUPAC Name: 3-methyl-4-nitrobenzonitrile
Synonyms: 3-methyl-4-nitrobenzonitrile
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: IHVNKSXTJZNBQA-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1[N+]([O-])=O)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results