missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-3-heptene (cis- and trans- mixture) 97.0+%, TCI America™
Supplier: TCI America M09915ML
Specifications
| 3-Methyl-3-heptene (cis- and trans- mixture) | |
| Colorless | |
| C8H16 | |
| 5 mL | |
| AAUHUDBDDBJONC-UHFFFAOYSA-N | |
| 3-methylhept-3-ene | |
| 138980 | |
| ≥97.0% (GC) |
| 7300-03-0 | |
| 121°C | |
| MFCD00048796 | |
| 3295 | |
| CCCC=C(C)CC | |
| 112.216 | |
| 112.22 | |
| Liquid |
Chemical Identifiers
| 7300-03-0 | |
| 112.216 | |
| AAUHUDBDDBJONC-UHFFFAOYSA-N | |
| 3-methylhept-3-ene |
| C8H16 | |
| MFCD00048796 | |
| 138980 | |
| CCCC=C(C)CC |
Safety and Handling
TSCA : No