missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-heptene (cis- and trans- mixture) 97.0+%, TCI America™

Catalog No. p-7132482
Change view
Click to view available options
Quantity:
1 mL
5 mL
Catalog No. Quantity
M09911ML 1 mL
M09915ML 5 mL
2 options

Catalog No. M09911ML

Supplier: TCI America M09911ML

Only null left

Chemical Identifiers

CAS 7300-03-0
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00048796
InChI Key AAUHUDBDDBJONC-UHFFFAOYSA-N
PubChem CID 138980
IUPAC Name 3-methylhept-3-ene
SMILES CCCC=C(C)CC

Specifications

Color Colorless
Boiling Point 121°C
Quantity 1 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-heptene (cis- and trans- mixture)