accessibility menu, dialog, popup

UserName

CH3H3CCH3(E)

CAS RN 7300-03-0

IUPAC Name: (3E)-3-methylhept-3-ene
Synonyms: (3E)-3-methylhept-3-ene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: AAUHUDBDDBJONC-BQYQJAHWSA-N
SMILES: CCC\C=C(/C)CC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results