missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-buten-1-ol 98.0+%, TCI America™

Supplier:  TCI America M0726100ML

 View more versions of this product

Catalog No. M0726100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Methyl-3-buten-1-ol
Yellow
C5H10O
100 mL
3-methyl-3-buten-1-ol, isobutenylcarbinol, 3-buten-1-ol, 3-methyl, isoprenol, methallylcarbinol, 3-isopentenyl alcohol, 2-methyl-1-buten-4-ol, isopropenylethyl alcohol, methallyl carbinol, 3-methyl-3-butenol
CC(=C)CCO
86.13
CHEBI:62898
≥98.0% (GC)
763-32-6
130°C
MFCD00002933
1987
CPJRRXSHAYUTGL-UHFFFAOYSA-N
3-methylbut-3-en-1-ol
12988
86.13
Liquid

Chemical Identifiers

763-32-6
86.13
CPJRRXSHAYUTGL-UHFFFAOYSA-N
12988
3-methylbut-3-en-1-ol
C5H10O
MFCD00002933
3-methyl-3-buten-1-ol, isobutenylcarbinol, 3-buten-1-ol, 3-methyl, isoprenol, methallylcarbinol, 3-isopentenyl alcohol, 2-methyl-1-buten-4-ol, isopropenylethyl alcohol, methallyl carbinol, 3-methyl-3-butenol
CHEBI:62898
CC(=C)CCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2393

RTECSNumber : EM9473100

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions