missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-buten-1-ol 98.0+%, TCI America™

Catalog No. p-7132323
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
M072625ML 25 mL
M0726100ML 100 mL
M0726500ML 500 mL
3 options

Catalog No. M072625ML

Supplier: TCI America M072625ML

Only null left

Chemical Identifiers

CAS 763-32-6
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00002933
InChI Key CPJRRXSHAYUTGL-UHFFFAOYSA-N
Synonym 3-methyl-3-buten-1-ol, isobutenylcarbinol, 3-buten-1-ol, 3-methyl, isoprenol, methallylcarbinol, 3-isopentenyl alcohol, 2-methyl-1-buten-4-ol, isopropenylethyl alcohol, methallyl carbinol, 3-methyl-3-butenol
PubChem CID 12988
ChEBI CHEBI:62898
IUPAC Name 3-methylbut-3-en-1-ol
SMILES CC(=C)CCO

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 mL
UN Number 1987
Formula Weight 86.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-buten-1-ol
EINECSNumber (2)-2393
RTECSNumber EM9473100
TSCA Yes