accessibility menu, dialog, popup

UserName

CAS RN 763-32-6

IUPAC Name: 3-methylbut-3-en-1-ol
Synonyms: 3-methyl-3-buten-1-ol isopentenyl alcohol 3-isopentenyl alcohol methallylcarbinol isobutenylcarbinol delta(3)-isopentenyl alcohol 3-isopentenol
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: CPJRRXSHAYUTGL-UHFFFAOYSA-N
SMILES: CC(=C)CCO
Boiling Point 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (5)