missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenol 98.0+%, TCI America™

Supplier:  TCI America M0122100ML

 View more versions of this product

Catalog No. M0122100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Methoxyphenol
Yellow
C7H8O2
100 mL
m-methoxyphenol, 3-hydroxyanisole, m-hydroxyanisole, phenol, 3-methoxy, m-guaiacol, resorcinol monomethyl ether, resorcinol methyl ether, 1-hydroxy-3-methoxybenzene, phenol, m-methoxy, 3-methoxy phenol
COC1=CC=CC(=C1)O
124.139
CHEBI:52678
≥98.0% (GC)
150-19-6
244°C
MFCD00002267
2810
ASHGTJPOSUFTGB-UHFFFAOYSA-N
3-methoxyphenol
9007
124.14
Liquid

Chemical Identifiers

150-19-6
124.139
ASHGTJPOSUFTGB-UHFFFAOYSA-N
9007
3-methoxyphenol
C7H8O2
MFCD00002267
m-methoxyphenol, 3-hydroxyanisole, m-hydroxyanisole, phenol, 3-methoxy, m-guaiacol, resorcinol monomethyl ether, resorcinol methyl ether, 1-hydroxy-3-methoxybenzene, phenol, m-methoxy, 3-methoxy phenol
CHEBI:52678
COC1=CC=CC(=C1)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0567

RTECSNumber : SL7524000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions