missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenol 98.0+%, TCI America™

Catalog No. p-7132004
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M012225ML 25 mL
M0122100ML 100 mL
M0122500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M012225ML Supplier TCI America Supplier No. M012225ML
Only null left

Chemical Identifiers

CAS 150-19-6
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00002267
InChI Key ASHGTJPOSUFTGB-UHFFFAOYSA-N
Synonym m-methoxyphenol, 3-hydroxyanisole, m-hydroxyanisole, phenol, 3-methoxy, m-guaiacol, resorcinol monomethyl ether, resorcinol methyl ether, 1-hydroxy-3-methoxybenzene, phenol, m-methoxy, 3-methoxy phenol
PubChem CID 9007
ChEBI CHEBI:52678
IUPAC Name 3-methoxyphenol
SMILES COC1=CC=CC(=C1)O

Specifications

Color Yellow
Boiling Point 244°C
Quantity 25 mL
UN Number 2810
Formula Weight 124.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxyphenol
EINECSNumber (3)-0567
RTECSNumber SL7524000
TSCA Yes