missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenethyl Alcohol 98.0+%, TCI America™

Catalog No. M26581G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26581G 1 g
M26585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26581G Supplier TCI America Supplier No. M26581G

Chemical Identifiers

CAS 5020-41-7
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00002893
InChI Key UPPGEJSCUZMCMW-UHFFFAOYSA-N
Synonym 2-3-methoxyphenyl ethanol, 3-methoxyphenethyl alcohol, benzeneethanol, 3-methoxy, 2-3-methoxyphenyl ethan-1-ol, phenethyl alcohol, m-methoxy, 3-methoxybenzeneethanol, m-methoxyphenethyl alcohol, 1-2-hydroxyethyl-3-methoxybenzene, acmc-20abd2, 3-methoxyphenethylalcohol
PubChem CID 78724
IUPAC Name 2-(3-methoxyphenyl)ethanol
SMILES COC1=CC=CC(=C1)CCO

Specifications

Color Yellow
Boiling Point 140°C
Quantity 1 g
Formula Weight 152.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxyphenethyl Alcohol