missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenethyl alcohol, 97%

Catalog No. p-4089696
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC182250010 1 g Glass bottle
1 options

Catalog No. AC182250010

Supplier: Thermo Scientific Chemicals 182250010

Only null left

CAS: 5020-41-7 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5020-41-7
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002893
InChI Key UPPGEJSCUZMCMW-UHFFFAOYSA-N
Synonym 2-3-methoxyphenyl ethanol, 3-methoxyphenethyl alcohol, benzeneethanol, 3-methoxy, 2-3-methoxyphenyl ethan-1-ol, phenethyl alcohol, m-methoxy, 3-methoxybenzeneethanol, m-methoxyphenethyl alcohol, 1-2-hydroxyethyl-3-methoxybenzene, acmc-20abd2, 3-methoxyphenethylalcohol
PubChem CID 78724
IUPAC Name 2-(3-methoxyphenyl)ethanol
SMILES COC1=CC=CC(=C1)CCO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0750g/mL
Boiling Point 141.0°C to 143.0°C (12.0 mmHg)
Flash Point >113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5381
Linear Formula CH3OC6H4CH2CH2OH
Quantity 1 g
Specific Gravity 1.075
Formula Weight 152.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methoxyphenethyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-705-2

RUO – Research Use Only