missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Methoxyphenacyl Bromide 98.0+%, TCI America™
Supplier: TCI America M081525G
Specifications
| 3′-Methoxyphenacyl Bromide | |
| 63°C | |
| C9H9BrO2 | |
| 25 g | |
| 2-bromo-1-3-methoxyphenyl ethanone, 2-bromo-3'-methoxyacetophenone, m-methoxyphenacyl bromide, 3'-methoxyphenacyl bromide, 3-bromoacetyl anisole, 2-bromo-1-3-methoxyphenyl ethan-1-one, 3-methoxyphenacyl bromide, bromomethyl 3-methoxyphenyl ketone, ccris 7998, 3-methoxyphenacylbromide | |
| COC1=CC=CC(=C1)C(=O)CBr | |
| 229.073 | |
| 229.07 | |
| Crystalline Powder |
| 5000-65-7 | |
| White-Yellow | |
| MFCD00000199 | |
| 3261 | |
| IOOHBIFQNQQUFI-UHFFFAOYSA-N | |
| 2-bromo-1-(3-methoxyphenyl)ethanone | |
| 101294 | |
| ≥98.0% (T) |
Chemical Identifiers
| 5000-65-7 | |
| 229.073 | |
| IOOHBIFQNQQUFI-UHFFFAOYSA-N | |
| 101294 | |
| COC1=CC=CC(=C1)C(=O)CBr |
| C9H9BrO2 | |
| MFCD00000199 | |
| 2-bromo-1-3-methoxyphenyl ethanone, 2-bromo-3'-methoxyacetophenone, m-methoxyphenacyl bromide, 3'-methoxyphenacyl bromide, 3-bromoacetyl anisole, 2-bromo-1-3-methoxyphenyl ethan-1-one, 3-methoxyphenacyl bromide, bromomethyl 3-methoxyphenyl ketone, ccris 7998, 3-methoxyphenacylbromide | |
| 2-bromo-1-(3-methoxyphenyl)ethanone |
Safety and Handling
TSCA : No