CAS RN 5000-65-7
2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific™
2-Bromo-3'-methoxyacetophenone, 98%
CAS: 5000-65-7 Molecular Formula: C9H9BrO2 Molecular Weight (g/mol): 229.073 MDL Number: MFCD00000199 InChI Key: IOOHBIFQNQQUFI-UHFFFAOYSA-N Synonym: 2-bromo-1-3-methoxyphenyl ethanone,2-bromo-3'-methoxyacetophenone,m-methoxyphenacyl bromide,3'-methoxyphenacyl bromide,3-bromoacetyl anisole,2-bromo-1-3-methoxyphenyl ethan-1-one,3-methoxyphenacyl bromide,bromomethyl 3-methoxyphenyl ketone,ccris 7998,3-methoxyphenacylbromide PubChem CID: 101294 IUPAC Name: 2-bromo-1-(3-methoxyphenyl)ethanone SMILES: COC1=CC=CC(=C1)C(=O)CBr
2-Bromo-3'-methoxyacetophenone, 97%
CAS: 5000-65-7 Molecular Formula: C9H9BrO2 Molecular Weight (g/mol): 229.07 MDL Number: MFCD00000199 InChI Key: IOOHBIFQNQQUFI-UHFFFAOYSA-N Synonym: 2-bromo-1-3-methoxyphenyl ethanone,2-bromo-3'-methoxyacetophenone,m-methoxyphenacyl bromide,3'-methoxyphenacyl bromide,3-bromoacetyl anisole,2-bromo-1-3-methoxyphenyl ethan-1-one,3-methoxyphenacyl bromide,bromomethyl 3-methoxyphenyl ketone,ccris 7998,3-methoxyphenacylbromide PubChem CID: 101294 IUPAC Name: 2-bromo-1-(3-methoxyphenyl)ethanone SMILES: COC1=CC=CC(=C1)C(=O)CBr
3'-Methoxyphenacyl Bromide 99.0+%, TCI America™
CAS: 5000-65-7 Molecular Formula: C9H9BrO2 Molecular Weight (g/mol): 229.073 MDL Number: MFCD00000199 InChI Key: IOOHBIFQNQQUFI-UHFFFAOYSA-N Synonym: 2-bromo-1-3-methoxyphenyl ethanone,2-bromo-3'-methoxyacetophenone,m-methoxyphenacyl bromide,3'-methoxyphenacyl bromide,3-bromoacetyl anisole,2-bromo-1-3-methoxyphenyl ethan-1-one,3-methoxyphenacyl bromide,bromomethyl 3-methoxyphenyl ketone,ccris 7998,3-methoxyphenacylbromide PubChem CID: 101294 IUPAC Name: 2-bromo-1-(3-methoxyphenyl)ethanone SMILES: COC1=CC=CC(=C1)C(=O)CBr
3'-Methoxyphenacyl Bromide 98.0+%, TCI America™
CAS: 5000-65-7 Molecular Formula: C9H9BrO2 Molecular Weight (g/mol): 229.073 MDL Number: MFCD00000199 InChI Key: IOOHBIFQNQQUFI-UHFFFAOYSA-N Synonym: 2-bromo-1-3-methoxyphenyl ethanone,2-bromo-3'-methoxyacetophenone,m-methoxyphenacyl bromide,3'-methoxyphenacyl bromide,3-bromoacetyl anisole,2-bromo-1-3-methoxyphenyl ethan-1-one,3-methoxyphenacyl bromide,bromomethyl 3-methoxyphenyl ketone,ccris 7998,3-methoxyphenacylbromide PubChem CID: 101294 IUPAC Name: 2-bromo-1-(3-methoxyphenyl)ethanone SMILES: COC1=CC=CC(=C1)C(=O)CBr