missing translation for 'onlineSavingsMsg'
Learn More

3'-Methoxyphenacyl Bromide 98.0+%, TCI America™

Catalog No. p-7132381
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08155G 5 g
M081525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08155G Supplier TCI America Supplier No. M08155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5000-65-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00000199
InChI Key IOOHBIFQNQQUFI-UHFFFAOYSA-N
Synonym 2-bromo-1-3-methoxyphenyl ethanone, 2-bromo-3'-methoxyacetophenone, m-methoxyphenacyl bromide, 3'-methoxyphenacyl bromide, 3-bromoacetyl anisole, 2-bromo-1-3-methoxyphenyl ethan-1-one, 3-methoxyphenacyl bromide, bromomethyl 3-methoxyphenyl ketone, ccris 7998, 3-methoxyphenacylbromide
PubChem CID 101294
IUPAC Name 2-bromo-1-(3-methoxyphenyl)ethanone
SMILES COC1=CC=CC(=C1)C(=O)CBr

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 229.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Methoxyphenacyl Bromide