missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzylmagnesium chloride, 0.25M solution in THF, AcroSeal™

Catalog No. AC431820500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431820500 50 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431820500 Supplier Thermo Scientific Chemicals Supplier No. 431820500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26905-40-8 | C8H9ClMgO | 180.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26905-40-8 , 109-99-9
Molecular Formula C8H9ClMgO
Molecular Weight (g/mol) 180.91
MDL Number MFCD01319896
InChI Key ZPCUEMLXJGELGI-UHFFFAOYSA-M
Synonym 3-methoxybenzylmagnesium chloride, 3-methoxybenzyl magnesiumchloride, 3-methoxybenzyl magnesium chloride, zpcuemlxjgelgi-uhfffaoysa-m, magnesium,chloro 3-methoxyphenyl methyl, 3-methoxybenzylmagnesium chloride 0.25 m in tetrahydrofuran, 3-methoxybenzylmagnesium chloride solution, 0.25 m in thf
PubChem CID 11263864
IUPAC Name chloro[(3-methoxyphenyl)methyl]magnesium
SMILES COC1=CC=CC(C[Mg]Cl)=C1

Specifications

CAS Min % 3.0, 93.0
CAS Max % 7.01, 97.0
Color Gray to Yellow
Density 0.9040g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3OC6H4CH2MgCl
Quantity 50 mL
Specific Gravity 0.904
Solubility Information Solubility in water: reacts
Formula Weight 180.92
Physical Form Solution
Chemical Name or Material 3-Methoxybenzylmagnesium chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only