accessibility menu, dialog, popup

UserName

CAS RN 26905-40-8

IUPAC Name: chloro[(3-methoxyphenyl)methyl]magnesium
Synonyms: chloro[(3-methoxyphenyl)methyl]magnesium
Molecular Weight (g/mol): 180.91
Molecular Formula: C8H9ClMgO
InChi Key: ZPCUEMLXJGELGI-UHFFFAOYSA-M
SMILES: COC1=CC=CC(C[Mg]Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results