missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxybenzylamine 98.0+%, TCI America™
Supplier: TCI America M085125ML
Specifications
| 3-Methoxybenzylamine | |
| Yellow | |
| C8H11NO | |
| 25 mL | |
| 3-methoxybenzylamine, m-methoxybenzylamine, 3-methoxyphenyl methanamine, 1-3-methoxyphenyl methanamine, benzenemethanamine, 3-methoxy, 3-methoxyphenyl methylamine, benzylamine, m-methoxy, methoxybenzylamine, 3, benzenemethanamine, 3-methoxy-9ci, sz4 | |
| COC1=CC=CC(CN)=C1 | |
| 137.18 | |
| 137.18 | |
| Liquid |
| 5071-96-5 | |
| 141°C | |
| MFCD00008115 | |
| 2735 | |
| GRRIMVWABNHKBX-UHFFFAOYSA-N | |
| 1-(3-methoxyphenyl)methanamine | |
| 21156 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 5071-96-5 | |
| 137.18 | |
| GRRIMVWABNHKBX-UHFFFAOYSA-N | |
| 21156 | |
| COC1=CC=CC(CN)=C1 |
| C8H11NO | |
| MFCD00008115 | |
| 3-methoxybenzylamine, m-methoxybenzylamine, 3-methoxyphenyl methanamine, 1-3-methoxyphenyl methanamine, benzenemethanamine, 3-methoxy, 3-methoxyphenyl methylamine, benzylamine, m-methoxy, methoxybenzylamine, 3, benzenemethanamine, 3-methoxy-9ci, sz4 | |
| 1-(3-methoxyphenyl)methanamine |
Safety and Handling
RTECSNumber : DP5550000
TSCA : No