missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzylamine 98.0+%, TCI America™

Catalog No. M08515ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08515ML 5 mL
M085125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08515ML Supplier TCI America Supplier No. M08515ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5071-96-5
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00008115
InChI Key GRRIMVWABNHKBX-UHFFFAOYSA-N
Synonym 3-methoxybenzylamine, m-methoxybenzylamine, 3-methoxyphenyl methanamine, 1-3-methoxyphenyl methanamine, benzenemethanamine, 3-methoxy, 3-methoxyphenyl methylamine, benzylamine, m-methoxy, methoxybenzylamine, 3, benzenemethanamine, 3-methoxy-9ci, sz4
PubChem CID 21156
IUPAC Name 1-(3-methoxyphenyl)methanamine
SMILES COC1=CC=CC(CN)=C1

Specifications

Color Yellow
Boiling Point 141°C
Quantity 5 mL
UN Number 2735
Formula Weight 137.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Methoxybenzylamine
RTECSNumber DP5550000
TSCA No