missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzylamine, 98%

Catalog No. AC170550100
Change view
Click to view available options
Quantity:
10 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170550100 10 mL Glass bottle
AC170550250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC170550100 Supplier Thermo Scientific Chemicals Supplier No. 170550100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5071-96-5 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5071-96-5
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00008115
InChI Key GRRIMVWABNHKBX-UHFFFAOYSA-N
Synonym 3-methoxybenzylamine, m-methoxybenzylamine, 3-methoxyphenyl methanamine, 1-3-methoxyphenyl methanamine, benzenemethanamine, 3-methoxy, 3-methoxyphenyl methylamine, benzylamine, m-methoxy, methoxybenzylamine, 3, benzenemethanamine, 3-methoxy-9ci, sz4
PubChem CID 21156
SMILES COC1=CC=CC(CN)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.0700g/mL
Boiling Point 140°C (37.0 mmHg)
Flash Point >110°C
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5470 to 1.549
Linear Formula CH3OC6H4CH2NH2
Quantity 10 mL
Beilstein 13, I, 226
Specific Gravity 1.07
IUPAC Name (3-methoxyphenyl)methanamine
Formula Weight 137.18
Percent Purity 98%
Chemical Name or Material 3-Methoxybenzylamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-779-6
RTECSNumber DP5550000
TSCA TSCA

RUO – Research Use Only