missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzyl chloride, 98+%

Catalog No. p-7020613
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1263014 25 g
AAA1263018 50 g
AAA1263030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1263014 Supplier Thermo Scientific Chemicals Supplier No. A1263014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 824-98-6 | C8H9ClO | 156.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 824-98-6
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00000907
InChI Key VGISFWWEOGVMED-UHFFFAOYSA-N
Synonym 3-methoxybenzyl chloride, 1-chloromethyl-3-methoxybenzene, m-methoxybenzyl chloride, m-chloromethyl anisole, benzene, 1-chloromethyl-3-methoxy, anisole, m-chloromethyl, 3-methoxybenzylchloride, 3-chloromethyl anisole, 3-methoxy benzyl chloride, benzyl chloride,3-methoxy
PubChem CID 69994
IUPAC Name 1-(chloromethyl)-3-methoxybenzene
SMILES COC1=CC=CC(CCl)=C1

Specifications

Density 1.078
Boiling Point 122°C to 123°C (12 mmHg)
Flash Point 101°C (213°F)
Refractive Index 1.544
Quantity 25 g
UN Number UN1760
Beilstein 636684
Sensitivity Moisture sensitive
Formula Weight 156.61
Percent Purity ≥98%
Chemical Name or Material 3-Methoxybenzyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 212-541-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only