missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzyl chloride, 97%

Catalog No. p-156855
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC197860050 5 g
AC197860250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC197860050 Supplier Thermo Scientific Chemicals Supplier No. 197860050
Only null left

CAS: 824-98-6 | C8H9ClO | 156.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 824-98-6
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00000907
InChI Key VGISFWWEOGVMED-UHFFFAOYSA-N
Synonym 3-methoxybenzyl chloride, 1-chloromethyl-3-methoxybenzene, m-methoxybenzyl chloride, m-chloromethyl anisole, benzene, 1-chloromethyl-3-methoxy, anisole, m-chloromethyl, 3-methoxybenzylchloride, 3-chloromethyl anisole, 3-methoxy benzyl chloride, benzyl chloride,3-methoxy
PubChem CID 69994
IUPAC Name 1-(chloromethyl)-3-methoxybenzene
SMILES COC1=CC=CC(CCl)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.0700g/mL
Boiling Point 124°C (13.0 mmHg)
Flash Point 101°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5430 to 1.545
Linear Formula CH3OC6H4CH2Cl
Quantity 5 g
Beilstein 06, I, 189
Specific Gravity 1.07
Formula Weight 156.61
Percent Purity 97%
Chemical Name or Material 3-Methoxybenzyl chloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 212-541-1
TSCA TSCA

RUO – Research Use Only