missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzotrifluoride 97.0+%, TCI America™

Catalog No. M22435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22435G 5 g
M224325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22435G Supplier TCI America Supplier No. M22435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 454-90-0
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.14
MDL Number MFCD00000385
InChI Key XHONYVFDZSPELQ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl anisole, 1-methoxy-3-trifluoromethyl benzene, 3-methoxybenzotrifluoride, 3-trifluoromethylanisole, benzene, 1-methoxy-3-trifluoromethyl, 3-methoxy-1-trifluoromethyl benzene, pubchem2712, 3-trifluoromethylanisol, acmc-1cuj0, m-trifluoromethyl anisole
PubChem CID 96691
IUPAC Name 1-methoxy-3-(trifluoromethyl)benzene
SMILES COC1=CC=CC(=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 160°C
Quantity 5 g
UN Number 1993
Formula Weight 176.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxybenzotrifluoride