CAS RN 454-90-0
IUPAC Name:
1-methoxy-3-(trifluoromethyl)benzene
Synonyms:
1-methoxy-3-(trifluoromethyl)benzene
Molecular Weight (g/mol):
176.14
Molecular Formula:
C8H7F3O
InChi Key:
XHONYVFDZSPELQ-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(=C1)C(F)(F)F
1
–
1
of
1
results
1
3-Methoxybenzotrifluoride 97.0+%, TCI America™
CAS: 454-90-0 Molecular Formula: C8H7F3O Molecular Weight (g/mol): 176.14 MDL Number: MFCD00000385 InChI Key: XHONYVFDZSPELQ-UHFFFAOYSA-N Synonym: 3-trifluoromethyl anisole,1-methoxy-3-trifluoromethyl benzene,3-methoxybenzotrifluoride,3-trifluoromethylanisole,benzene, 1-methoxy-3-trifluoromethyl,3-methoxy-1-trifluoromethyl benzene,pubchem2712,3-trifluoromethylanisol,acmc-1cuj0,m-trifluoromethyl anisole PubChem CID: 96691 IUPAC Name: 1-methoxy-3-(trifluoromethyl)benzene SMILES: COC1=CC=CC(=C1)C(F)(F)F