missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxybenzenethiol 98.0+%, TCI America™
Supplier: TCI America M119425G
Specifications
| 3-Methoxybenzenethiol | |
| Yellow | |
| C7H8OS | |
| 25 g | |
| 3-methoxythiophenol, 3-methoxybenzenthiol, 3-methoxy thiophenol, m-methoxythiophenol, 3-mercaptoanisole, benzenethiol, 3-methoxy, m-methoxybenzenethiol, 3-methoxy-benzenethiol, 3-methoxybenzene-1-thiol, meta-methoxybenzenethiol | |
| COC1=CC(=CC=C1)S | |
| 140.2 | |
| 140.20 | |
| Liquid |
| 15570-12-4 | |
| 114°C | |
| MFCD00004841 | |
| 2810 | |
| QMVAZEHZOPDGHA-UHFFFAOYSA-N | |
| 3-methoxybenzenethiol | |
| 84989 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 15570-12-4 | |
| 140.2 | |
| QMVAZEHZOPDGHA-UHFFFAOYSA-N | |
| 84989 | |
| COC1=CC(=CC=C1)S |
| C7H8OS | |
| MFCD00004841 | |
| 3-methoxythiophenol, 3-methoxybenzenthiol, 3-methoxy thiophenol, m-methoxythiophenol, 3-mercaptoanisole, benzenethiol, 3-methoxy, m-methoxybenzenethiol, 3-methoxy-benzenethiol, 3-methoxybenzene-1-thiol, meta-methoxybenzenethiol | |
| 3-methoxybenzenethiol |
Safety and Handling
TSCA : No