missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzenethiol 98.0+%, TCI America™

Catalog No. p-7132589
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11945G 5 g
M119425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11945G Supplier TCI America Supplier No. M11945G
Only null left

Chemical Identifiers

CAS 15570-12-4
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00004841
InChI Key QMVAZEHZOPDGHA-UHFFFAOYSA-N
Synonym 3-methoxythiophenol, 3-methoxybenzenthiol, 3-methoxy thiophenol, m-methoxythiophenol, 3-mercaptoanisole, benzenethiol, 3-methoxy, m-methoxybenzenethiol, 3-methoxy-benzenethiol, 3-methoxybenzene-1-thiol, meta-methoxybenzenethiol
PubChem CID 84989
IUPAC Name 3-methoxybenzenethiol
SMILES COC1=CC(=CC=C1)S

Specifications

Color Yellow
Boiling Point 114°C
Quantity 5 g
UN Number 2810
Formula Weight 140.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxybenzenethiol