missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-N,N-dimethylaniline, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0870009

 View more versions of this product

Catalog No. AAL0870009

Only null left
Explore available promotions

Description

Description

N,N-Dimethyl-m-anisidine is a phenyl amide with antineoplastic and antibacterial properties. It can be used to produce C20H19NO4. It will need reagent tributyl orthoformate and solvent propan-2-ol with reaction time of 90 min.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-m-anisidine is a phenyl amide with antineoplastic and antibacterial properties. It can be used to produce C20H19NO4. It will need reagent tributyl orthoformate and solvent propan-2-ol with reaction time of 90 min.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

3-Methoxy-N,N-dimethylaniline
1.031
C9H13NO
MFCD00051779
UN2810
3-dimethylaminoanisole, n,n-dimethyl-m-anisidine, 3-methoxy-n,n-dimethylbenzenamine, 3-methoxyphenyl dimethylamine, 3-dimethylamino-anisole, acmc-1c3gj, m-methoxy-n,n-dimethylanilin, n,n-dimethyl-3-methoxyaniline, n1,n1-dimethyl-3-methoxyaniline, 3-methoxy-n,n-dimethylbenzeneamine
MOYHVSKDHLMMPS-UHFFFAOYSA-N
3-methoxy-N,N-dimethylaniline
139977
98%
15799-79-8
113°C to 114°C (6 mmHg)
1.559
10 g
1423591
Not miscible or difficult to mix in water.
CN(C)C1=CC(=CC=C1)OC
151.209
151.21

Chemical Identifiers

15799-79-8
151.209
MOYHVSKDHLMMPS-UHFFFAOYSA-N
139977
CN(C)C1=CC(=CC=C1)OC
C9H13NO
MFCD00051779
3-dimethylaminoanisole, n,n-dimethyl-m-anisidine, 3-methoxy-n,n-dimethylbenzenamine, 3-methoxyphenyl dimethylamine, 3-dimethylamino-anisole, acmc-1c3gj, m-methoxy-n,n-dimethylanilin, n,n-dimethyl-3-methoxyaniline, n1,n1-dimethyl-3-methoxyaniline, 3-methoxy-n,n-dimethylbenzeneamine
3-methoxy-N,N-dimethylaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only