missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-5-(trifluoromethyl)aniline 98.0+%, TCI America™

Catalog No. M22625G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22625G 5 g
M22621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22625G Supplier TCI America Supplier No. M22625G

Chemical Identifiers

CAS 349-55-3
Molecular Formula C8H8F3NO
Molecular Weight (g/mol) 191.153
MDL Number MFCD00000393
InChI Key VTFGJEYZCUWSAM-UHFFFAOYSA-N
Synonym 3-methoxy-5-trifluoromethyl aniline, 3-amino-5-methoxybenzotrifluoride, 5-trifluoromethyl-m-anisidine, benzenamine, 3-methoxy-5-trifluoromethyl, 3-trifluoromethyl-5-methoxybenzenamine, 3-methoxy-5-trifluoromethyl-phenylamine, 3-methoxy-5-trifluoromethyl benzenamine, 5-methoxy-alpha,alpha,alpha-trifluoro-m-toluidine, 5-methoxy-3-trifluoromethyl phenylamine
PubChem CID 605671
IUPAC Name 3-methoxy-5-(trifluoromethyl)aniline
SMILES COC1=CC(=CC(=C1)N)C(F)(F)F

Specifications

Melting Point 52°C
Quantity 5 g
Formula Weight 191.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)aniline