CAS RN 349-55-3
IUPAC Name:
3-methoxy-5-(trifluoromethyl)aniline
Synonyms:
3-methoxy-5-(trifluoromethyl)aniline
Molecular Weight (g/mol):
191.15
Molecular Formula:
C8H8F3NO
InChi Key:
VTFGJEYZCUWSAM-UHFFFAOYSA-N
SMILES:
COC1=CC(N)=CC(=C1)C(F)(F)F
1
–
1
of
1
results
1
3-Methoxy-5-(trifluoromethyl)aniline 98.0+%, TCI America™
CAS: 349-55-3 Molecular Formula: C8H8F3NO Molecular Weight (g/mol): 191.153 MDL Number: MFCD00000393 InChI Key: VTFGJEYZCUWSAM-UHFFFAOYSA-N Synonym: 3-methoxy-5-trifluoromethyl aniline,3-amino-5-methoxybenzotrifluoride,5-trifluoromethyl-m-anisidine,benzenamine, 3-methoxy-5-trifluoromethyl,3-trifluoromethyl-5-methoxybenzenamine,3-methoxy-5-trifluoromethyl-phenylamine,3-methoxy-5-trifluoromethyl benzenamine,5-methoxy-alpha,alpha,alpha-trifluoro-m-toluidine,5-methoxy-3-trifluoromethyl phenylamine PubChem CID: 605671 IUPAC Name: 3-methoxy-5-(trifluoromethyl)aniline SMILES: COC1=CC(=CC(=C1)N)C(F)(F)F