Learn More
3-Methoxy-4-nitrobenzoic acid, 98+%
CAS: 5081-36-7 | C8H7NO5 | 197.146 g/mol
Supplier: Thermo Scientific Chemicals L0889114
Description
3-Methoxy-4-nitrobenzoic acid was used in detection of nitroaromatic compounds by CdSe- quantum dots capped with polyamidoamine dendrimer-1,12-diaminododecane core-generation 4 nanocomposites. It was used in the synthesis of vorozole derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methoxy-4-nitrobenzoic acid was used in detection of nitroaromatic compounds by CdSe- quantum dots capped with polyamidoamine dendrimer-1,12-diaminododecane core-generation 4 nanocomposites. It was used in the synthesis of vorozole derivatives.
Solubility
Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 3-Methoxy-4-nitrobenzoic acid | |
| 231°C to 235°C | |
| MFCD00007353 | |
| 2696042 | |
| Insoluble in water. | |
| COC1=C(C=CC(=C1)C(=O)O)[N+](=O)[O-] | |
| 78764 | |
| ≥98% |
| 5081-36-7 | |
| C8H7NO5 | |
| 25 g | |
| 3-methoxy-4-nitrobenzoic acid, 4-nitro-3-methoxybenzoic acid, 4-nitro-m-anisic acid, benzoic acid, 3-methoxy-4-nitro, 3-methoxy-4-nitrobenzoicacid, pubchem3918, acmc-1bn9s, ksc270m6t, 3-methoxy4-nitro-benzoic acid, 3-methoxy-4-nitro-benzoic acid | |
| PWURRRRGLCVBMX-UHFFFAOYSA-N | |
| 197.146 | |
| 197.15 |
Chemical Identifiers
| 5081-36-7 | |
| 197.146 | |
| PWURRRRGLCVBMX-UHFFFAOYSA-N | |
| 78764 |
| C8H7NO5 | |
| MFCD00007353 | |
| 3-methoxy-4-nitrobenzoic acid, 4-nitro-3-methoxybenzoic acid, 4-nitro-m-anisic acid, benzoic acid, 3-methoxy-4-nitro, 3-methoxy-4-nitrobenzoicacid, pubchem3918, acmc-1bn9s, ksc270m6t, 3-methoxy4-nitro-benzoic acid, 3-methoxy-4-nitro-benzoic acid | |
| COC1=C(C=CC(=C1)C(=O)O)[N+](=O)[O-] |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 225-793-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only